Anti-HIF1A Mouse Monoclonal Antibody (Fluorescein) [clone: 241812]

Supplier: R & D Systems
IC1935F
220018-260EA 438.18 USD
220018-260
Anti-HIF1A Mouse Monoclonal Antibody (Fluorescein) [clone: 241812]
Antibodies
Type: Primary
Antigen: HIF1A
Clonality: Monoclonal
Clone: 241812
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR