Anti-ABCC1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: QCRL]

Supplier: R & D Systems
IC19291P
220018-258EA 513.38 USD
220018-258
Anti-ABCC1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: QCRL]
Antibodies
Type: Primary
Antigen: ABCC1
Clonality: Monoclonal
Clone: QCRL
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR