Anti-SOX17 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 614013]

Supplier: R & D Systems
IC19241G
102884-402EA 561.11 USD
102884-402
Anti-SOX17 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 614013]
Antibodies
Type: Primary
Antigen: SOX17
Clonality: Monoclonal
Clone: 614013
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR