Anti-FN1 Mouse Monoclonal Antibody (Fluorescein) [clone: P1H11]

Supplier: R & D Systems
IC1918F
220018-254EA 420.83 USD
220018-254
Anti-FN1 Mouse Monoclonal Antibody (Fluorescein) [clone: P1H11]
Antibodies
Type: Primary
Antigen Symbol: FN1
Antigen Name: fibronectin 1
Antigen Synonyms: CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
Clonality: Monoclonal
Clone: P1H11
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR