Anti-IL23A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 320244]

Supplier: R & D Systems
IC18871P
102884-400EA 513.38 USD
102884-400
Anti-IL23A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 320244]
Antibodies
Type: Primary
Antigen: IL23A
Clonality: Monoclonal
Clone: 320244
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR