Anti-TGF-beta 1/2/3 Mouse Monoclonal Antibody (PE) [clone: 1D11]

Supplier: R & D Systems
IC1835P
220018-251EA 577.02 USD
220018-251
Anti-TGF-beta 1/2/3 Mouse Monoclonal Antibody (PE) [clone: 1D11]
Antibodies
Type: Primary
Antigen Symbol: TGF-beta 1/2/3
Antigen Name:
Antigen Synonyms:
Clonality: Monoclonal
Clone: 1D11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR