Anti-IL27 Rat Monoclonal Antibody (Fluorescein) [clone: 234205]

Supplier: R & D Systems
IC1834F
220018-249EA 420.83 USD
220018-249
Anti-IL27 Rat Monoclonal Antibody (Fluorescein) [clone: 234205]
Antibodies
Type: Primary
Antigen: IL27
Clonality: Monoclonal
Clone: 234205
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR