Anti-EBI3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 355022]

Supplier: R & D Systems
IC18341P
102884-390EA 549.54 USD
102884-390
Anti-EBI3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 355022]
Antibodies
Type: Primary
Antigen: EBI3
Clonality: Monoclonal
Clone: 355022
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR