Anti-EBI3 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 355022]

Supplier: R & D Systems
IC18341A
102884-386EA 614.62 USD
102884-386
Anti-EBI3 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 355022]
Antibodies
Type: Primary
Antigen: EBI3
Clonality: Monoclonal
Clone: 355022
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR