Anti-STAT3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 232209]

Supplier: R & D Systems
IC1799P
220018-248EA 501.82 USD
220018-248
Anti-STAT3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 232209]
Antibodies
Type: Primary
Antigen: STAT3
Clonality: Monoclonal
Clone: 232209
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR