Anti-GATA1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 234732]

Supplier: R & D Systems
IC1779P
220018-246EA 513.38 USD
220018-246
Anti-GATA1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 234732]
Antibodies
Type: Primary
Antigen Symbol: GATA1
Antigen Name: GATA binding protein 1 (globin transcription factor 1)
Antigen Synonyms: ERYF1, GATA-1, GF-1, GF1, NF-E1, NFE1, XLANP, XLTDA, XLTT
Clonality: Monoclonal
Clone: 234732
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR