Anti-HMGB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 115603]

Supplier: R & D Systems
IC1690A
102884-376EA 552.43 USD
102884-376
Anti-HMGB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 115603]
Antibodies
Type: Primary
Antigen: HMGB1
Clonality: Monoclonal
Clone: 115603
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR