Anti-STAT1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 246523]

Supplier: R & D Systems
IC1490G
102884-370EA 561.11 USD
102884-370
Anti-STAT1 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 246523]
Antibodies
Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone: 246523
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR