Anti-STAT1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 246523]

Supplier: R & D Systems
IC1490A
102884-368EA 594.37 USD
102884-368
Anti-STAT1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 246523]
Antibodies
Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone: 246523
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR