Anti-AIFM1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
IC1457G
102884-364EA 552.43 USD
102884-364
Anti-AIFM1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: AIFM1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR