Anti-ALB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 188835]

Supplier: R & D Systems
IC1455P
102884-362EA 513.38 USD
102884-362
Anti-ALB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 188835]
Antibodies
Type: Primary
Antigen: ALB
Clonality: Monoclonal
Clone: 188835
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR