Anti-SPP1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 223126]

Supplier: R & D Systems
IC14331P
102884-356EA 543.75 USD
102884-356
Anti-SPP1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 223126]
Antibodies
Type: Primary
Antigen: SPP1
Clonality: Monoclonal
Clone: 223126
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR