Anti-SPP1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 223126]

Supplier: R & D Systems
IC14331A
102884-352EA 577.02 USD
102884-352
Anti-SPP1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 223126]
Antibodies
Type: Primary
Antigen: SPP1
Clonality: Monoclonal
Clone: 223126
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR