Anti-ADAM10 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1633]

Supplier: R & D Systems
IC1427A
220018-233EA 594.37 USD
220018-233
Anti-ADAM10 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1633]
Antibodies
Type: Primary
Antigen: ADAM10
Clonality: Monoclonal
Clone: 1633
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR