Anti-ACTA2 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 1A4]

Supplier: R & D Systems
IC1420C
102884-346EA 642.09 USD
102884-346
Anti-ACTA2 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 1A4]
Antibodies
Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone: 1A4
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR