Anti-BGLAP Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 190125]

Supplier: R & D Systems
IC1419P
220018-230EA 480.12 USD
220018-230
Anti-BGLAP Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 190125]
Antibodies
Type: Primary
Antigen: BGLAP
Clonality: Monoclonal
Clone: 190125
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR