Anti-BGLAP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 190125]

Supplier: R & D Systems
IC1419A
102884-340EA 513.38 USD
102884-340
Anti-BGLAP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 190125]
Antibodies
Type: Primary
Antigen: BGLAP
Clonality: Monoclonal
Clone: 190125
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR