Anti-INS Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 182410]

Supplier: R & D Systems
IC1417A
220018-229EA 655.11 USD
220018-229
Anti-INS Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 182410]
Antibodies
Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: 182410
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR