Anti-IL25 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 207702]

Supplier: R & D Systems
IC13991P
220018-228EA 543.75 USD
220018-228
Anti-IL25 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 207702]
Antibodies
Type: Primary
Antigen: IL25
Clonality: Monoclonal
Clone: 207702
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR