Anti-CDH2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 401408]

Supplier: R & D Systems
IC1388P
102884-338EA 485.91 USD
102884-338
Anti-CDH2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 401408]
Antibodies
Type: Primary
Antigen: CDH2
Clonality: Monoclonal
Clone: 401408
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR