Anti-IL26 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 510414]

Supplier: R & D Systems
IC13751A
220018-223EA 577.02 USD
220018-223
Anti-IL26 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 510414]
Antibodies
Type: Primary
Antigen: IL26
Clonality: Monoclonal
Clone: 510414
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR