Anti-TNFSF13B Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 121808]

Supplier: R & D Systems
IC1357A
102884-330EA 577.02 USD
102884-330
Anti-TNFSF13B Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 121808]
Antibodies
Type: Primary
Antigen: TNFSF13B
Clonality: Monoclonal
Clone: 121808
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR