Anti-TP53 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 184727]

Supplier: R & D Systems
IC13551A
220018-221EA 608.83 USD
220018-221
Anti-TP53 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 184727]
Antibodies
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: 184727
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR