Anti-CTNNB1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 196624]

Supplier: R & D Systems
IC13292P
220018-216EA 501.82 USD
220018-216
Anti-CTNNB1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 196624]
Antibodies
Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 196624
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR