Anti-CTNNB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 196624]

Supplier: R & D Systems
IC13292A
220018-214EA 608.83 USD
220018-214
Anti-CTNNB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 196624]
Antibodies
Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 196624
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR