Anti-NES Mouse Monoclonal Antibody (APC) [clone: 196908]

Supplier: R & D Systems
IC1259A
220018-211EA 594.37 USD
220018-211
Anti-NES Mouse Monoclonal Antibody (APC) [clone: 196908]
Antibodies
Type: Primary
Antigen Symbol: NES
Antigen Name: nestin
Antigen Synonyms: Nbla00170
Clonality: Monoclonal
Clone: 196908
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR