Anti-IL25 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 182203]

Supplier: R & D Systems
IC1258P
220018-210EA 501.82 USD
220018-210
Anti-IL25 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 182203]
Antibodies
Type: Primary
Antigen: IL25
Clonality: Monoclonal
Clone: 182203
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR