Anti-IL25 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 182203]

Supplier: R & D Systems
IC1258A
220018-208EA 584.25 USD
220018-208
Anti-IL25 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 182203]
Antibodies
Type: Primary
Antigen: IL25
Clonality: Monoclonal
Clone: 182203
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR