Anti-SEMA3A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 215803]

Supplier: R & D Systems
IC1250P
102884-274EA 543.75 USD
102884-274
Anti-SEMA3A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 215803]
Antibodies
Type: Primary
Antigen: SEMA3A
Clonality: Monoclonal
Clone: 215803
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR