Anti-TNFSF13B Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 137314]

Supplier: R & D Systems
IC1241P
220018-204EA 513.38 USD
220018-204
Anti-TNFSF13B Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 137314]
Antibodies
Type: Primary
Antigen: TNFSF13B
Clonality: Monoclonal
Clone: 137314
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR