Anti-beta-III Tubulin Mouse Monoclonal Antibody [clone: TUJ1] (PerCP (Peridinin-Chlorophyll Protein Complex))

Supplier: R & D Systems
IC1195C
220018-199EA 642.09 USD
220018-199
Anti-beta-III Tubulin Mouse Monoclonal Antibody [clone: TUJ1] (PerCP (Peridinin-Chlorophyll Protein Complex))
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: beta-III Tubulin
Antigen Synonyms: Tubulin Beta 3, Tubulin beta-4
Clonality: Monoclonal
Clone: TUJ1
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR