Anti-6xHis Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: AD1.1.1]

Supplier: R & D Systems
IC050G
102884-062EA 600.15 USD
102884-062
Anti-6xHis Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: AD1.1.1]
Antibodies
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: AD1.1.1
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR