Anti-DIG Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 611621]

Supplier: R & D Systems
HAM7520
102884-126EA 561.11 USD
102884-126
Anti-DIG Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 611621]
Antibodies
Type: Primary
Antigen: DIG
Clonality: Monoclonal
Clone: 611621
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR