Anti-pY Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 179003]

Supplier: R & D Systems
HAM1676
220018-111EA 566.89 USD
220018-111
Anti-pY Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 179003]
Antibodies
Type: Primary
Antigen: pY
Clonality: Monoclonal
Clone: 179003
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR