Anti-TRP53 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: R & D Systems
HAF1355
220018-110EA 485.91 USD
220018-110
Anti-TRP53 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: TRP53
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR