Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: R & D Systems
HAF109
220018-109EA 229.94 USD
220018-109
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Simple Western

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR