Anti-IgG Goat Polyclonal Antibody

Supplier: R & D Systems
G-202-C
220018-100EA 623.29 USD
220018-100
Anti-IgG Goat Polyclonal Antibody
Antibodies
Hidden

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR