Anti-FITC Goat Polyclonal Antibody

Supplier: R & D Systems
G-148-C
220018-098EA 633.42 USD
220018-098
Anti-FITC Goat Polyclonal Antibody
Antibodies
Hidden

Type: Primary
Antigen Symbol: FITC
Antigen Name: Fluorescein Isothiocyanate
Antigen Synonyms:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR