Anti-Hemoglobin Goat Polyclonal Antibody

Supplier: R & D Systems
G-134-C
220018-097EA 633.42 USD
220018-097
Anti-Hemoglobin Goat Polyclonal Antibody
Antibodies
Hidden

Type: Primary
Antigen Symbol:
Antigen Name: Hemoglobin
Antigen Synonyms:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR