Anti-TNNI3 (Peptide 3) Goat Polyclonal Antibody

Supplier: R & D Systems
G-129-C
220018-096EA 616.06 USD
220018-096
Anti-TNNI3 (Peptide 3) Goat Polyclonal Antibody
Antibodies
Hidden

Type: Primary
Antigen: TNNI3 (Peptide 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR