Anti-IgE Goat Polyclonal Antibody

Supplier: R & D Systems
G-107-C
220018-093EA 623.29 USD
220018-093
Anti-IgE Goat Polyclonal Antibody
Antibodies
Hidden

Type: Primary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR