Anti-IgM µ Goat Polyclonal Antibody

Supplier: R & D Systems
G-106-C-ABS
220018-092EA 623.29 USD
220018-092
Anti-IgM µ Goat Polyclonal Antibody
Antibodies
Hidden

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR