Anti-IgG Goat Polyclonal Antibody

Supplier: R & D Systems
G-101-C-ABS
220018-089EA 163.42 USD
220018-089
Anti-IgG Goat Polyclonal Antibody
Antibodies
Hidden

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR