Anti-VEGF R1, R2, R3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 89106, 49560, 54733]

Supplier: R & D Systems
FABSP3P
220018-051EA 485.91 USD
220018-051
Anti-VEGF R1, R2, R3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 89106, 49560, 54733]
Antibodies
Type: Primary
Antigen: VEGF R1, R2, R3
Clonality: Monoclonal
Clone: 89106, 49560, 54733
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR