Anti-SELPLG Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 688101]

Supplier: R & D Systems
FAB9961P
102884-104EA 480.12 USD
102884-104
Anti-SELPLG Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 688101]
Antibodies
Type: Primary
Antigen: SELPLG
Clonality: Monoclonal
Clone: 688101
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR